Learn More
Invitrogen™ CADM4 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518838
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
Specifications
| CADM4 | |
| Polyclonal | |
| Unconjugated | |
| CADM4 | |
| Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 199731 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8NFZ8 | |
| CADM4 | |
| Synthetic peptide sequence (REAFLNGSDGHKRKE) corresponding to the C-terminus amino acids of CADM4 (aa 370-385). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |