Learn More
SGE™ GC Septa: CS
Click to view available options
Diameter (English) Inner:
0.18 in.
0.37 in.
0.39 in.
0.43 in.
0.45 in.
0.5 in.
0.59 in.
Unit Size:
Pack of 48
Pack of 50
Description
CS Septa
- PTFE-coated silicone septa for low temperature applications
- Good durability, good resealing
- Excellent solvent resistance; good tear resistance
- Maximum temperature: 200°C (392°F)
Specifications
Specifications
| Diameter (English) Inner | 0.37 in. |
| Material | PTFE-coated Silicone |
| Quantity | 50/Pk. |