Learn More
Applied Biosystems™ TaqMan™ TAMRA Probe
Click to view available options
Quantity:
20,000 pmol
50,000 pmol
6,000 pmol
Product Type:
TAMRA Probe
Unit Size:
Each
Description
Custom TaqMan™ probes are dual-labeled primers suitable for use in any real-time PCR applicaton. They incorporate a 5' reporter dye and a 3' TAMRA™ nonfluorescent quencher. Choose among FAM™, VIC™, ABY™, and JUN™ as 3' reporters. Probes are HPLC-purified.
Specifications
Specifications
| 3'Primer Modification | TAMRA |
| 5'Primer Modification | FAM, TET, VIC |
| Content And Storage | 1 tube containing a dual-labeled probe in solution or dried down. Store at -15°C to -25°C |
| Form | Liquid or Dried Down |
| Format | Tube |
| For Use With (Equipment) | 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex |
| Green Features | Sustainable packaging |
| Internal Probe Modification | None |
| Product Line | TaqMan |
| Product Type | TAMRA Probe |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur