Learn More
Sartorius™ Grade 292 High Purity Folded Filter Qualitative Filter Papers
Shop All Sartorius Products
Click to view available options
Diameter (Metric):
90 mm
110 mm
125 mm
150 mm
185 mm
240 mm
270 mm
385 mm
Unit Size:
Pack of 100
Description
- Made of refined pulp and cotton linters with an >95% α-cellulose content,
- Ash content <0.1% according to DIN 54370
- Wet-strengthened
- Supplied in folded format
Specifications
Specifications
| Format | Folded |
| Material | Cotton Linters, Refined Pulp |
| Particle Retention | 5 to 8 μm |
| Filtration Speed | 60μm/sec. |
| For Use With (Application) | Used for analytical purposes that require a low ash content |
| Grade | 292 |
| Quantity | 100/Pk. |
| Thickness (Metric) | 0.18 mm |
| Type | Qualitative Filter Paper |
| Diameter (Metric) | 125 mm |