Learn More
Macherey Nagel HPLC COLUMN EC NUCLEOSIL 50-5 C8ec endcappedUSP phase L7, eluent in columnMeCN/H2O
Brand: Macherey Nagel 720092.46
HPLC COLUMN EC NUCLEOSIL 50-5 C8ec endcappedUSP phase L7, eluent in columnMeCN/H2O