Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 40
Description
- Free of detectable DNase/RNase, human DNA and pyrogens
- Contents non-cytotoxic
- Alphanumeric well coding
- High clarity of the microplates for optimal microscopic examinations
- Stackable
- Consecutive lot numbering
- Uniform footprint; conforms to (ANSI) (ANSI/SBS 1-2004)
- Risk of contamination from sample material being carried over is minimised
- μClear™ bottom extends detection capability down to 340nm
- Well profile: F-bottom/chimney well (flat bottom)
- Growth area per well: 34 mm²
- Working volume per well: 25 μl - 340 μl
- Sterile
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Well Shape | Flat Bottom (F-well) |
| Quantity | 40/Cs. |
| Show More |