Learn More
Greiner Bio-One μClear™ 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
20/Pk., 120/Cs.
8/Pk.
Unit Size:
Pack of 120
Pack of 32
Beskrivelse
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic, non-cytotoxic
- Compatible with automated systems
- Stackable
- Alphanumeric well coding μClear™ bottom
- Well profile: F-bottom (flat bottom)
- Growth area per well: 10 mm²
- Working volume per well: 15 μl - 110 μl
- Available with lid that enables gas exchange with the lowest possible evaporation
- Sterile
Tekniske data
Tekniske data
| Barcode | Alphanumeric |
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Packaging | 8/Pk. |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion