Learn More
Azlon™ PVDF and Polyethylene Stopcock
Click to view available options
Connectors:
2-way
3-way
Material:
PVDF
Polyethylene
For Use With (Equipment):
6 - 7mm I.D. tube
6 to 7mm I.D. tube
8 - 9mm I.D tube
8 to 9mm I.D. tube
Unit Size:
Each
Description
- PVDF has excellent temperature, chemical and ageing resistance
- Polyethylene available for less stringent requirements
Specifications
Specifications
| Material | Polyethylene |
| Autoclavable | Not Autoclavable |
| No. per Pack | 1 |
| For Use With (Equipment) | 6 to 7mm I.D. tube |
| Connectors | 3-way |