missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vidarabine Phosphate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Vidarabine Phosphate
Specifications
Specifications
| Chemical Name or Material | Vidarabine Phosphate |
| CAS | 29984-33-6 |
| Analyte or Component Names | Vidarabine Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H14 N5 O7 P |
| Certifications/Compliance | ISO 17025 |
| Synonym | 9H-Purin-6-amine, 9-(5-O-phosphono-β-D-arabinofuranosyl)-, Adenine, 9-β-D-arabinofuranosyl-, 5'-(dihydrogen phosphate) (8CI), 9-(5-O-Phosphono-β-D-arabinofuranosyl)-9H-purin-6-amine, 5'-Arabinosyladenine monophosphate, 9-β-D-Arabinofuranosyl AMP, 9-β-D-Arabinofuranosyladenine 5'-monophosphate, 9-β-D-Arabinofuranosyladenine 5'-phosphate, 9-β-D-Arabinofuranosyladenine monophosphate, Adenine arabinonucleoside 5'-phosphate, Adenine arabinoside 5'-monophosphate, Adenine arabinoside monophosphate, Arabinosyladenine monophosphate, CI 808, D-Arabinosyladenine 5'-monophosphate, NSC 127223, Vidarabine monophosphate, Vidarabine phosphate, Vidarabine-5'-monophosphate, ara-AMP |
| InChI Formula | InChI=1S/C10H14N5O7P/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(22-10)1-21-23(18,19)20/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20)/t4-,6-,7+,10-/m1/s1 |
| SMILES | Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)(O)O)[C@@H](O)[C@@H]3O |
| IUPAC Name | [(2R,3S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction