missing translation for 'onlineSavingsMsg'
Learn More

Victoria blue B, MP Biomedicals™

Product Code. 11469771 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469771 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11469771 Supplier MP Biomedicals Supplier No. 0215829125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2580-56-5
Molecular Formula C33H32ClN3
Molecular Weight (g/mol) 506.09
InChI Key LLWJPGAKXJBKKA-UHFFFAOYSA-N
Synonym Basic Blue B, Basic Blue 26
PubChem CID 17407
ChEBI CHEBI:87651
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]naphthalen-1-ylidene]-phenylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[NH+]C3=CC=CC=C3)C4=CC=CC=C24)C5=CC=C(C=C5)N(C)C.[Cl-]

Specifications

Formula Weight 506.1
Color Blue to Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Victoria blue B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.