missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vicriviroc maleate, MedChemExpress
Description
Vicriviroc maleate (SCH-417690 maleate
Specifications
Specifications
| Chemical Name or Material | Vicriviroc maleate |
| CAS | 599179-03-0 |
| Color | Light Nude |
| Purity Grade Notes | Research |
| Molecular Formula | C32H42F3N5O6 |
| Quantity | 5 mg |
| Synonym | SCH-417690 maleateSCH-D maleate |
| Solubility Information | DMSO : 50 mg/mL (76.96 mM; Need ultrasonic) ∣H2O : 25 mg/mL (38.48 mM; Need ultrasonic and warming) |
| SMILES | CC1(N2C[C@H](C)N([C@H](C3=CC=C(C(F)(F)F)C=C3)COC)CC2)CCN(C(C4=C(C)N=CN=C4C)=O)CC1.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 649.7 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?