missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Jour Chromatography™ Polypropylene Flanged Fingertight Nuts, 1/16 in.
Click to view available options
Color:
Black
White
Unit Size:
Pack of 10
Description
- Compatible to all 1/4"-28 flat bottom ports
- For 1/16" OD tubing
- Fiber glass reinforced PP with PP washers
Specifications
Specifications
| Type | Nut |
| Color | Black |
| Packaging Quantity | 10 Pack |
| For Use With | For 1.5 mm Tubing |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction