missing translation for 'onlineSavingsMsg'
Learn More

VICI Jour Chromatography™ Polypropylene Flanged Fingertight Nuts, 1/16 in.

Product Code. 12985021
Click to view available options
Color:
Black
White
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12985021

Brand: VICI Jour Chromatography™ JR201094BK10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For flanged connections

  • Compatible to all 1/4"-28 flat bottom ports
  • For 1/16" OD tubing
  • Fiber glass reinforced PP with PP washers

Specifications

Type Nut
Color Black
Packaging Quantity 10 Pack
For Use With For 1.5 mm Tubing
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.