missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Jour Chromatography™ PEEK Fingertight Nut
Click to view available options
For Use With:
1/16 in. Tubing
Unit Size:
Pack of 5
Specifications
Specifications
| Type | Nut |
| Color | Green |
| Packaging Quantity | 5 Pack |
| Thread Size | 10 to 32 |
| For Use With | 1/16 in. Tubing |
| Material | PEEK |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction