missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Jour Chromatography™ ETFE Tubing
ETFE (ethyltrifluoroethylene) tubing is particularly suitable for HPLC, since it has a higher burst pressure as compared to PTFE.
Brand: VICI Jour Chromatography™ JR-T-082-M5
Code : 16
Additional Details : Weight : 1.00000kg
Description
- Ideal polymer for applications with chloride salts, which typically corrode stainless steel
Specifications
Tubing | |
1.59 mm | |
ETFE |
0.5 mm | |
5 m |