missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vibostolimab Humanized Recombinant Human Monoclonal Antibody
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Vibostolimab is a biosimilar that targets TIGIT. Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications
Specifications
| Antigen | Vibostolimab Humanized |
| Applications | ELISA, SDS-Page |
| Classification | Recombinant Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | MK-7684 |
| Host Species | Human |
| Immunogen | Human TIGIT/VSIG9/VSTM3. |
| Purification Method | Protein A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?