Learn More
Thermo Scientific™ Vial Kit with Polyseal Caps
Product Code. 12418626
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Product Code. 12418626
Brand: Thermo Scientific™ B78012PS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample.
  • Clear vial
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Silicone
Color Clear
Volume (Metric) 4 mL
Quantity 100/Pk.