missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter™ Vi-CELL™ Quad Pak Reagent Kit with Vials

Product Code. 13402309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
220mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13402309 1 Kit 220mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13402309 Supplier Beckman Coulter™ Supplier No. 383198

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with the Beckman Coulter Vi-CELL™ XR cell viability analyzer. Beckman Coulter™ Vi-CELL™ Quad Pak Reagent Kit with Vials includes 4 individual Vi-CELL™ particle reagent kits and 4 bags of sample vials (120 per bag).

Reagent Pak contains:

  • Trypan blue
  • Buffer solution
  • Disinfectant
  • Cleaning agent

Applications: Bioprocessing, Cell Analysis, Cell Health, Cell Line Development, Cell Cycle

Specifications

Chemical Name or Material Particle reagent kits
Quantity 1 Kit
Formulation Tryptan blue, buffer, disinfectant, cleaning agents
Includes 4 x 220 mL buffer, disinfectant, cleaning; 4 x 110 mL Typtan blue
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vi-CELL Quad and Single Pak
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 1
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P370+P378-In case of fire: Use water spray to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P233-Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl Alcohol; Trypan Blue, Sodium Azide; Diazolidinyl Urea, Subtilisin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.