missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VHR/DUSP3 Polyclonal specifically detects VHR/DUSP3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | VHR/DUSP3 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | dual specificity phosphatase 3, dual specificity protein phosphatase 3, Dual specificity protein phosphatase VHR, EC 3.1.3.16, EC 3.1.3.48, Vaccinia H1-related phosphatase, vaccinia virus phosphatase VH1-related, VHRserine/threonine specific protein phosphatase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant VHR/DUSP3 (P51452; Met1-Pro185). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?