missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VHL Monoclonal Antibody (Ig33), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513940
This item is not returnable.
View return policy
Description
MA5-13940 targets von Hippel-Lindau Protein in immunohistochemistry (paraffin) and immunofluorescence applications and shows reactivity with Human and mouse samples. The MA5-13940 immunogen is a recombinant protein containing aa 1-213 of human VHL.
Von Hippel-Lindau syndrome (VHL) is a dominantly inherited familial cancer syndrome predisposing to a variety of malignant and benign tumors. A germline mutation of this gene is the basis of familial inheritance of VHL syndrome. The protein encoded by this gene is a component of the protein complex that includes elongin B, elongin C, and cullin-2, and possesses ubiquitin ligase E3 activity. This protein is involved in the ubiquitination and degradation of hypoxia-inducible-factor (HIF), which is a transcription factor that plays a central role in the regulation of gene expression by oxygen. RNA polymerase II subunit POLR2G/RPB7 is also reported to be a target of this protein. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| VHL | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P40337, P40338 | |
| VHL | |
| A recombinant protein containing aa 1-213 of human VHL | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Ig33 | |
| Unconjugated | |
| VHL | |
| elongin binding protein; HRCA1; Protein G7; pVHL; RCA1; VHL; VHL1; VHL19; Vhlh; VHLp18(MEA); Von Hippel Lindau; von hippel-lindau; von Hippel-Lindau disease tumor suppressor; von Hippel-Lindau syndrome homolog; von Hippel-Lindau syndrome protein homolog; von Hippel-Lindau tumor suppressor; von Hippel-Lindau tumor suppressor, E3 ubiquitin protein ligase | |
| Mouse | |
| Protein G | |
| RUO | |
| 22346, 7428 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction