missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ VH 101 phenol-alkylC6-amine

Product Code. 18798176
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18798176 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18798176 Supplier R&D Systems™ Supplier No. 7294/25
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. VHL ligand with alkyl linker and terminal amine for onward chemistry

VH 101 phenol-alkylC6-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC6 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,4 R)-N-(2-((6-Aminohexyl)oxy)-4-(4-methylthiazol-5-yl)benzyl)-1-(-2-(1-fluorocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide dihydrochloride
CAS 2564467-16-7
Quantity 25 mg
Product Line Tocris
Target Degrader Building Blocks
Molecular Formula C32H46FN5O5S0.2 HCl
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.