missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 032, amine
GREENER_CHOICE

Product Code. 18724759
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724759 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724759 Supplier Tocris Bioscience™ Supplier No. 6462/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Functionalized VHL ligand for PROTACs

VH 032, amine is a derivative of the von Hippel-Lindau (VHL) ligand, VH 032; commonly used as a precursor to a PROTAC® that hijacks VHL as the E3 ubiquitin ligase component. Supplied with a primary amine functional handle at a position known not to significantly affect binding to VHL, for ready conjugation to a linker/target protein ligand.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Require a different derivative? Get in touch

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4 R)-3-Methyl-L-valyl-4-hydroxy-N-[[4-(4-methyl-5-thiazolyl)phenyl]methyl]-L-prolinamide dihydrochloride
CAS 2137142-47-1
Synonym (4R)-3-Methyl-L-valyl-4-hydroxy-N-[[4-(4-methyl-5-thiazolyl)phenyl]methyl]-L-prolinamide hydrochloride
Quantity 100 mg
Target Degrader Building Blocks
Molecular Formula C22H30N4O3S0.2 HCl
Formula Weight 467.02
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.