missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 032 amide-PEG3-BCN

Product Code. 30040310
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040310 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040310 Supplier Tocris Bioscience™ Supplier No. 8123/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Functionalized VHL building block for copper-free click chemistry

VH 032 amide-PEG3-BCN is a functionalized von Hippel Lindau (VHL) protein ligand for PROTAC™ research and development. Incorporates an E3 ligase ligand plus a PEG3 linker with terminal bicyclooctyne (BCN). It can be used for copper-free strain-promoted azide-alkyne cycloaddition (SPAAC) reaction. Part of a range of functionalized tool molecules for PROTAC R&D.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ((1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-yl)methyl ((S)-14-((2S,4R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidine-1-carbonyl)-15,15-dimethyl-12-oxo-3,6,9-trioxa-13-azahexadecyl)carbamate
CAS 2876198-36-4
Quantity 10 mg
Target Degrader Building Blocks
Molecular Formula C42H59N5O9S
Purity 0.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.