missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 032 amide-alkylC3-amine

Product Code. 18736506
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18736506 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18736506 Supplier Tocris Bioscience™ Supplier No. 7073/25
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

VHL ligand with alkyl linker and terminal amine for onward chemistry

VH 032 amide-alkylC3-amine is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkyl C3 linker and terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-(4-Aminobutanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride
CAS 2564467-25-8
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C26H37N5O4S0.2 HCl
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.