missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 032 amide-alkylC2-amine
GREENER_CHOICE

Product Code. 18737606
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 18737606

Brand: Tocris Bioscience™ 6852/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VH 032-AlkylC2-Amine

VH 032 amide-alkylC2-amine is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

This product has been recently renamed. The previous name for this product was VH 032 - linker 7

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4R)-1-((S)-2-(3-Aminopropanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride
Synonym VH 032 - linker 7
Quantity 25 mg
Molecular Formula C25H35N5O4S.2HCl
Formula Weight 574.56
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.