missing translation for 'onlineSavingsMsg'
Learn More

VGX-1027, MedChemExpress

Product Code. 16402677
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16402677

Brand: Medchem Express HY15507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VGX-1027 is an orally active isoxazole compound that exhibits various immunomodulatory properties. VGX-1027 targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, IL-10. VGX-1027 has antidiabetogenic effects by limiting cytokine-mediated immunoinflammatory events.

VGX-1027 is an orally active isoxazole compound that exhibits various immunomodulatory properties. VGX-1027 targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, IL-10. VGX-1027 has antidiabetogenic effects by limiting cytokine-mediated immunoinflammatory events.

Specifications

Chemical Name or Material VGX-1027
Physical Form Solid
CAS 6501-72-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H11NO3
For Use With (Application) COVID-19-immunoregulation
Synonym GIT 27
Solubility Information DMSO : ≥ 56 mg/mL (272.89 mM)
SMILES O=C(O)CC1CC(C2=CC=CC=C2)=NO1
Molecular Weight (g/mol) 205.21
Formula Weight 205.21
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.