missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25261-1-AP-20UL
This item is not returnable.
View return policy
Description
VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.Specifications
| VGLUT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A6 | |
| DNPI, SLC17A6, VGLUT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140919, 57084 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BLE7, Q9P2U8 | |
| SLC17A6 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction