missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VGLUT2 Monoclonal Antibody (N29/29), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545510
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45510 was sufficient for detection of VGLut2 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60kDa. This antibody was formerly sold as clone S29-29.
VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.
Specifications
| VGLUT2 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q8BLE7, Q9JI12, Q9P2U8 | |
| SLC17A6 | |
| Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N29/29 | |
| PE | |
| SLC17A6 | |
| 2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 140919, 57084, 84487 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction