Learn More
Invitrogen™ VGLUT1 Monoclonal Antibody (N28/9), PerCP
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545505
Description
1 μg/mL of MA5-45505 was sufficient for detection of VGLut1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 52kDa. No cross-reactivity against VGlut2. This antibody was formerly sold as clone S28-9.
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
Specifications
| VGLUT1 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q3TXX4, Q62634, Q9P2U7 | |
| SLC17A7 | |
| Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| N28/9 | |
| PerCP | |
| SLC17A7 | |
| 2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 116638, 57030, 72961 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.