missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VGLUT1 Monoclonal Antibody (A7A3)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA544957

Product Code. 17827852

  • £419.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VGLUT1 Monoclonal Antibody for Western Blot

A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VGLUT1
Monoclonal
2 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q3TXX4, Q62634, Q9P2U7
SLC17A7
Synthetic peptide within Human VGluT1 aa 501-550/560.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG3
Western Blot
A7A3
Unconjugated
SLC17A7
2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1
Mouse
Protein G
RUO
116638, 57030, 72961
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.