missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT1 Antibody (S28-9), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59329IR
This item is not returnable.
View return policy
Description
VGLUT1 Monoclonal specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VGLUT1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SLC17A7 | |
| Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1 | |
| 100 μL | |
| 57030 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| S28-9 | |
| Western Blot, Immunohistochemistry | |
| BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects 52kDa. No cross-reactivity against VGlut2. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction