missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLL2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84327-0.1ML
This item is not returnable.
View return policy
Description
VGLL2 Polyclonal specifically detects VGLL2 in Mouse samples. It is validated for Western Blot.
Specifications
| VGLL2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 245806 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Protein VITO1, transcription cofactor vestigial-like 2, transcription cofactor vestigial-like protein 2, vestigial like 2 (Drosophila), vgl-2, VGL2, VITO1vestigial-like 2 | |
| The immunogen is a synthetic peptide corresponding to a region of Mouse VGLL2 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur