missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10124-2-AP-20UL
This item is not returnable.
View return policy
Description
May act as a specific coactivator for the mammalian TEFs.Specifications
| VGLL1 | |
| Polyclonal | |
| Unconjugated | |
| VGLL1 | |
| Protein TONDU, TDU, vestigial like 1 (Drosophila), Vgl 1, VGL1, VGLL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170828, 363508, 51442 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99990, Q99NC0 | |
| VGLL1 | |
| VGLL1 Fusion Protein Ag0169 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction