missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VGLL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568954
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%.
May act as a specific coactivator for the mammalian TEFs.
Specifications
| VGLL1 | |
| Polyclonal | |
| Unconjugated | |
| VGLL1 | |
| Protein TONDU; RGD1564038; TDU; TONDU; Transcription cofactor vestigial-like protein 1; vestigial like 1; vestigial like 1 homolog; vestigial like 1 homolog (Drosophila); vestigial like family member 1; vestigial-like family member 1; vestigial-related factor; VGL1; Vgl-1; VGLL1; WUGSC:H_GS188P18.1b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170828 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99NC0 | |
| VGLL1 | |
| synthetic peptide directed towards the C-terminal region of mouse VGLL1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction