Learn More
VGAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14471-1-AP-150UL
 View more versions of this product
Product Code. 16883124
  • £349.00 / 150µL
Description
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Specifications
Specifications
VGAT
Polyclonal
Unconjugated
SLC32A1
GABA and glycine transporter, hVIAAT, SLC32A1, Vesicular GABA transporter, VGAT, VIAAT
Rabbit
Antigen Affinity Chromatography
RUO
140679, 22348, 83612
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35458, O35633, Q9H598
Slc32a1
VGAT Fusion Protein Ag5843
150 μL
Primary
Human, Mouse, Rat