Learn More
Invitrogen™ VGAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596231
Description
Positive Samples: Mouse spinal cord, Mouse brain; Cellular Location: Cytoplasmic vesicle membrane, Multi-pass membrane protein Immunogen sequence: MATLLRSKLS NVATSVSNKS QAKMSGMFAR MGFQAATDEE AVGFAHCDDL DFEHRQGLQM DILKAEGEPC GDEGAEAPVE GDIHYQRGSG APLPPSGSKD QVGGGGEFGG HDKPKITAWE AGWNVTNAIQ GMFVLGLPYA ILHGGYLGLF LIIFAAVVCC YTGKILIACL YEENEDGEVV RVRDSYVAIA NACCAPRFPT LGGRVVNVAQ IIELVMTCIL YVVVSGNLMY NSFPGLPVSQ KSWSIIATAV LLPCAFLKNL KAVSKFSLLC TLAHFVINIL VIAYCLSRAR DWAWEKVKFY IDVKKFPISI GIIVFSYTSQ IFLPSLEGNM QQPSEFHCMM NWTHIAACVL KGLFALVAYL TWADETKEVI TDNLPGSIRA VVNIFLVAKA LLSYPLPFFA AVEVLEKSLF QEGSRAFFPA CYSGDGRLKS WGLTLRCALV VFTLLMAIYV PHFALLMGLT GSLTGAGLCF LLPSLFHLRL LWRKLLWHQV FFDVAIFVIG GICSVSGFVH SLEGLIEAYR TNAED.
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Specifications
| VGAT | |
| Polyclonal | |
| Unconjugated | |
| SLC32A1 | |
| GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140679, 22348, 83612 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35458, O35633, Q9H598 | |
| SLC32A1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-525 of human SLC32A1 (Q9H598). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.