missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VGAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527569
This item is not returnable.
View return policy
Description
Recommended positive controls: SLC32A1-transfected 293T. Predicted reactivity: Mouse (97%), Rat (95%), Xenopus laevis (83%), Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Specifications
| VGAT | |
| Polyclonal | |
| Unconjugated | |
| SLC32A1 | |
| GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140679, 22348, 83612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.52 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O35458, O35633, Q9H598 | |
| SLC32A1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human VGAT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction