missing translation for 'onlineSavingsMsg'
Learn More

VG5Q Antibody (368011) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30732799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732799 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732799 Supplier Novus Biologicals Supplier No. FAB30481J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

VG5Q Monoclonal antibody specifically detects VG5Q in Mouse samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen VG5Q
Applications Immunocytochemistry
Classification Monoclonal
Clone 368011
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein
Host Species Rat
Immunogen E. coli-derived recombinant mouse VG5Q, Met1-Glu711, Accession # Q7TN31
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 55109
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.