missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Four gel trays (Length: 70, 100, 150, 200cm)
- Trays with aluminum casting gates
- Multiple comb slots to position along gel tray
- Color coded combs
- Buffer recirculation ports
- Coloured loading strips for easy well detection when loading
- Compact tank to reduce buffer volume required
- UV-transparent acrylic casting tray
Specifications
Gel Electrophoresis Unit | |
Vari-Gel | |
15 | |
19 x 31.5 x 7 cm | |
150 mm |
200 x 150 mm | |
4 | |
950 mL | |
200 mm | |
75 to 150 V |