missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas VG-FAST96 High Throughput Vari-gel Maxi System
Click to view available options
Dimensions (L x W):
17.5 x 16 cm
Unit Size:
Each
Description
- Single gel tray with fluorescent numbering, allows up to 96 SSPs to be loaded directly onto the tray
- Six comb slots
- Six 17 sample combs; each marker lane positioned centrally
- Compact tank to reduce buffer voulme required to cover gel, over voltage gradient and run-time
- Robust acrylic casting tray; UV transparent for visualisation with fluorescent numbering for SNP
- Used with 96 well microtitre plates, or 96 well blocks
- Buffer recirculation ports to connect to peristaltic pump for buffer recirculation during electrophoresis
Specifications
Specifications
| Type | Gel Electrophoresis Unit |
| Includes | Lid and cables, gel holder with cover plate 6 and 1mm 17-well combs |
| Material | Acrylic |
| Sample Size | 96 + 8 |
| Volume (Metric) | 950 mL |
| Volume (Metric) Buffer | 950 mL |
| Dimensions (L x W) | 17.5 x 16 cm |
| Dimensions (L x W x H) | 19.5 x 33 x 7.5 cm |
| Voltage | 100 to 150 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
