missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™-Plus qPCR Master Mix

Product Code. 30057773 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 Reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30057773 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057773 Supplier Applied Biosystems™ Supplier No. 4415327
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with VetMAX™ Reagents and VetMAX™ Controls.

The VetMAX™-Plus qPCR Master Mix is designed for multiplex amplification of animal pathogen DNA targets using a rapid qPCR strategy. The kit includes Xeno™ DNA Control, which serves as an internal positive control for DNA purification and qPCR. DNA is amplified using AmpliTaq Gold™ DNA Polymerase on a real-time PCR system.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Includes 100 reactions with the following components: qPCR Master Mix, Xeno™ DNA Control, and Nuclease-free water.
Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance Low
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System
Passive Reference Dye ROX (Pre-mixed)
Product Line VetMAX
Product Type PCR Reagents and Detection Kit for Animal Health
Quantity 100 Reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Sample Type DNA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.