missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™-Plus Multiplex One Step RT-PCR Kit

Product Code. 30057789 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30057789 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057789 Supplier Applied Biosystems™ Supplier No. 4415330
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with VetMAX™ Reagents and VetMAX™ Controls.

p>The VetMAX™-Plus Multiplex One-Step RT-PCR Kit is designed for multiplex amplification of animal pathogen RNA targets using a rapid, single-tube, real-time, reverse transcription PCR (RT-PCR) strategy. The kit includes Xeno™ RNA Control, which serves as an internal positive control for RNA purification and RT-PCR amplification. RNA is reverse-transcribed into cDNA, and then your target is amplified using AmpliTaq Gold™ DNA Polymerase on a real-time PCR system.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Includes 100 reactions with the following components: Multiplex RT-PCR Buffer, Multiplex RT-PCR Enzyme Mix, Xeno™ RNA Control, and Nuclease-free water.
Detection Method Primer-probe
Form Liquid
Format Tube
GC-Rich PCR Performance Low
PCR Method 1-step RT-qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System
Passive Reference Dye ROX (Pre-mixed)
Product Line VetMAX
Product Type PCR Reagents and Detection Kit for Animal Health
Quantity 100 reactions
Reverse Transcriptase ArrayScript™ UP
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Sample Type RNA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.