Learn More
Honeywell™ Vertigo White PU 3 Gloves
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
Description
- White Spectra™ / Lycra™ knitted glove
- White polyurethane coating on the palm and fingertips
- Elastic wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The cool Spectra™ touch gives an added sensation of comfort
- The glove's ventilated back optimises its breathability
- Thread provide outstanding properties against cuts
- Polyurethane coating at the palm and fingertips for outstanding abrasion resistance
- Caution! Do not expose this glove to heat
- White color enhances its hygiene and cleanliness properties
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2003 Mechanical hazards |
| Description | 13 gauge |
| Color | White |
| Material | Polyurethane, Lycra |
| Size | 6 |
| Length (Metric) | 24 cm |