missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Vertical Profile Auto-Desiccator Cabinet

Product Code. 10046511 Shop All SP Scienceware Products
Click to view available options
Color:
Clear with Blue End Caps
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10046511

Brand: Bel-Art™ SP Scienceware™ F420741226

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with these Auto-Desiccator Cabinets

  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Automatic dehumidification and desiccant regeneration cycle operates continuously every 20 minutes
  • Color-indicating silica gel inside the desiccant regeneration module allows quick visual confirmation of desiccant status
  • Large door with easy-to-view digital hygrometer maximizes access to interior space; includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens right to left
  • Perforated shelves facilitate circulation of dry air
  • Compact 34.1W x 41.4cmD footprint for space-saving stacking up to 2 units high

Specifications

Color Clear with Blue End Caps
Cabinet Style Auto-Desiccator
Material Copolyester
No. of Shelves 3
No. per Pack 1 Each
Shelf Type Perforated
Width (Metric) 34.1 cm
Height (Metric) 51.8 cm
Depth (Metric) 41.4 cm
Voltage 230 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.