Learn More
Scientific Industries SI™ Vertical Microtube Holder
Description
- Holds tubes vertically
- Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
- Optimized for high speed vortexing a small number of tubes
- No additional attachment necessary
Specifications
Specifications
| For Use With (Equipment) | Vortex-Genie 2 Mixer and Vortex-Genie Pulse |
| No. of Positions | 24 |