Learn More
Wesbart™ Vertical Cold Stores Racks
Product Code. 16202471
Click to view available options
:
160 x 150 x 630 mm
Product Code. 16202471
Brand: Wesbart™ VC561/9LB
Only null left
Add to basket
Add to basket
Wesbart™ Vertical Racks made of aluminum or stainless steel, available in a range of sizes and capacities for standard cryobox storage in chest freezers
  • Vertical racks for standard cryobox storage in chest freezers
  • Available in aluminum or stainless steel
  • Range of sizes and capacities
  • Solid sides for strength and rigidity
  • Fitted with a fold-down top handle for easy rack removal
Specifications
Type Freezer Storage Rack
For Use With (Equipment) Upright Freezers (Labcold ULTF301)
Includes Top Handle
Dimensions (L x W x H) Exterior 160 x 150 x 630 mm
Holds Boxes (8)