missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Verti-Gel Mini Tetrad PAGE System
Durable and leakproof system to run up to four gels.
£520.00 - £725.00
Specifications
Type | Verti-Gel Mini Tetrad PAGE System |
---|---|
Gel Dimensions | 75 x 80 mm |
Material | Injection-molded |
No. of Gels | 4 |
Thickness (Metric) | 0.5 mm, 0.75 mm, 1 mm, 1.5 mm, 2 mm combs |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15136624
|
Fisherbrand™
|
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 0.75mm bonded spacers, four notched plates, four 12-well 0.75mm combs |
£520.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
15156624
|
Fisherbrand™
|
100 x 100mm blotting module, caster and external stand, four notched/plain plates with 0.75mm spacer, four plate, four 12-well 0.75mm comb, four cassettes, eight pads |
£725.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
15176624
|
Fisherbrand™
|
100 x 100mm blotting module, caster and external stand, four notched/plain plates with 1.5mm spacer, four plates, four 12-well 1.5mm combs, four cassettes, eight pads |
£725.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
15116624
|
Fisherbrand™
|
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 1mm bonded spacers, four notched plates, four 12-well 1mm combs |
£520.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
- Injection-moulded construction
- Compatible with all 80 x 100mm and 100 x 100mm precast gels
- Low buffer volumes
Specifications
Verti-Gel Mini Tetrad PAGE System | |
Injection-molded | |
0.5 mm, 0.75 mm, 1 mm, 1.5 mm, 2 mm combs | |
190 x 130 x 150 mm |
75 x 80 mm | |
4 | |
250 to 1200 mL |