missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verteporfin, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Verteporfin
Specifications
Specifications
| Chemical Name or Material | Verteporfin |
| CAS | 129497-78-5 |
| Analyte or Component Names | Verteporfin |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C41 H42 N4 O8 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 23H,25H-Benzo[b]porphine-9,13-dipropanoic acid, 18-ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-, monomethyl ester, trans-, BPD-MA, CL 318952, Verteporfin, Visudyne, (4R,4aS)-rel-18-Ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-benzo[b]porphine-9,13-dipropanoic acid monomethyl ester |
| InChI Formula | InChI=1S/C41H42N4O8/c1-9-23-20(2)29-17-34-27-13-10-26(39(49)52-7)38(40(50)53-8)41(27,5)35(45-34)19-30-22(4)25(12-15-37(48)51-6)33(44-30)18-32-24(11-14-36(46)47)21(3)28(43-32)16-31(23)42-29/h9-10,13,16-19,38,43,45H,1,11-12,14-15H2,2-8H3,(H,46,47)/b31-16-,33-18-,34-17-,35-19-/t38-,41+/m1/s1 |
| SMILES | CC1=C(C=C)/C(N/C1=C\C2=N/C([C@@]3(C)C2=CC=C(C(OC)=O)[C@H]3C(OC)=O)=C\4)=C/C5=N/C(C(CCC(O)=O)=C5C)=C\C6=C(CCC(OC)=O)C(C)=C4N6 |
| IUPAC Name | 3-[rac-(23R,24S)-22,23-bis(methoxycarbonyl)-5-(3-methoxy-3-oxo-propyl)-4,10,15,24-tetramethyl-14-vinyl-25,26,27,28-tetrazahexacyclo[16.6.1.13,6.18,11.113,16.019,24]octacosa-1,3(28),4,6,8,10,12,14,16(26),17,19,21-dodecaen-9-yl]propanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction