missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Verteporfin

Product Code. 17159981
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 17159981

Brand: STEMCELL Technologies 1000261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

YAP1 inhibitor

  • Verteporfin is a benzoporphyrin derivative that inhibits autophagosome formation by directly targeting and modifying p62, a scaffold protein that targets polyubiquitinated proteins for degradation (Donohue et al.) Verteporfin also disrupts YAP-TEAD interactions by enhancing trypsin cleavage of YAP (EC50 = 100 nM; Brodowska et al.; Liu-Chittenden et al.)
  • Inhibits the growth, proliferation, and viability of human retinoblastoma cells (Brodowska et al.)
  • Inhibits self-renewal in cancer stem cells with high YAP1 and ALDH1 levels (Song et al.)

Specifications

CAS 129497-78-5
Synonym BPD-MA; CL 318,952
Quantity 1 mg
Molecular Formula C41H42N4O8
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.