missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Verso 1-Step RT-PCR Kits

Artikelnummer. 11842113
Click to view available options
Description:
Master Mix with Thermo-Start Taq (Hot Start)
Master Mix with ThermoPrime Taq
ReddyMix Master Mix with ThermoPrime Taq
Quantity:
400 x 25µL Rxns
80 x 25µL Rxns
Packungsgröße:
40 reactions
400 reactions
80 reactions
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11842113

Marke: Thermo Scientific™ AB1454/A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Get everything needed for RT-PCR in this complete set of reagents with a highly processive reverse transcriptase enzyme, available with regular or hot start Taq.

Verso 1-Step RT-PCR system is a complete set of RT-PCR reagents that combines everything needed for RT-PCR. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures. Verso also shows a better dynamic range than standard AMV or M-MuLV enzymes, with increases in template concentration giving rise to corresponding increases in product generated over a wider range of starting material concentrations.

The Verso system is available with ThermoPrime (standard PCR) or Thermo-Start (hot start) polymerase, the latter providing a more specific option that delivers higher yields and greatly reduces nonspecific primer extension before PCR. Thermo-Start Taq DNA Polymerase is a chemically-modified polymerase that remains inactive throughout the RT step, activated only by the high-temperature incubation (95°C for 15 minutes) prior to PCR.

An optional RT Enhancer is provided with kits and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment.

The Verso 1-step RT-PCR system is also available with a ReddyMix option, where the master mix contains a dye and precipitant that allow RT-PCR reactions to be loaded directly onto an electrophoresis gel. The red dye migrates between bromophenol blue and xylene cyanol at approximately 300 bp, depending on agarose concentration.

Highlights

  • Eliminate the need for DNase treatments -RT Enhancer (optional) prevents gDNA carry-over
  • Enzyme choice -Standard PCR or hot start options available
  • Available as ReddyMix formulation -eliminate post-PCR steps required for electrophoresis
  • RT and PCR steps performed in single sealed tube -convenience and reduced contamination risk
  • Verso enzyme is highly sensitive in a broad dynamic range -accurately detect RNA input from 1 pg to 1 µg
  • Wide working temperature range (42 to57°C) for success with GC-rich and other difficult templates
  • RNase Inhibitor included in RT enzyme mix
TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage Store at -20°C. Avoid freeze/thaw cycles
Description Master Mix with ThermoPrime Taq
Product Type Verso 1-Step RT-PCR Master Mix w/ThermoPrime Taq
Quantity 80 x 25µL Rxns
For Use With (Application) RT-PCR

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt